Logic® Vue Class II Enclosures, for Protection and Containment for Large Instruments
Lieferant: LABCONCO
3300411
LBCP3300411EA 0 EUR
LBCP3300411
Logic® Vue Class II Enclosures, for Protection and Containment for Large Instruments
Dunstabzugshauben und Verkleidungen Biologische Sicherheitswerkbänke
The Logic® Vue is a specialised class II enclosure providing personnel and product protection for large automated instruments such as liquid handlers. The Logic® Vue provides laminar ISO 5 air.
  • Large capacity enclosure
  • Accommodates diverse range of instruments such as liquid handlers and cell sorters
  • Provides accessibility from front and back
  • Visibility from all sides
  • Powered by MyLogic™ OS to interface with a variety of devices for additional researcher safety
Standards: Meets NSF/ANSI standard 49.
Verpackung: The international packaging indicated for selected units includes reduced height for high cube shipping containers.
Bitte beachten Installation required.