Sash-glider kit
Lieferant: LABCONCO
3408500
LBCP3408500EA 0 EUR
LBCP3408500
Sash-glider kit
Wischtücher Reinigungskits und -Pads
Use the Sash-glider to clean both sides of a biological safety cabinet's sash while it remains closed, to avoid contamination risks.
Lieferumfang: Sash-glider kit includes magnetic outside and inside blades, 2 microfibre sheets and 10 disposable white scrubbing strips. Additional scrubbing strips are available.