Biological safety cabinets, class II, type A2, Purifier® Cell Logic®+
Lieferant: LABCONCO
322691150 342591130
LBCP322691150EA 0 EUR
LBCP322691150 LBCP342591130
Biological safety cabinets, class II, type A2, Purifier® Cell Logic®+
Dunstabzugshauben und Verkleidungen Biologische Sicherheitswerkbänke
These Biosafety cabinets provide personnel, product and environmental protection from hazardous particulates such as agents that require Biosafety Level 1, 2 or 3 containment. Other appropriate applications include work with antineoplastic drugs, genetic material, asbestos and additional substances that generate hazardous airborne particulates. All models are NSF compliant. Cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional canopy connection. When canopy-connected, the biosafety cabinets may be used for applications involving minute quantities of volatile toxic chemicals and tracer amounts of radionuclides as an adjunct to microbiological research.
  • Excels in user comfort and functionality
  • Ergonomic design: Inclination™ Technology Suite that includes the following: 10° angled sash, patented curved air grille with built in armrest, fully accessible controls and eye-level mounted colour display with MyLogic™ operating system
  • Provides intuitive programming, colour coded menus, status messages and alarms with diagnostic troubleshooting