Equipment table
Lieferant: LABCONCO
3819100
LBCP3819100EA 0 EUR
LBCP3819100
Equipment table
Möbel Tische und Werkbänke
This mobile equipment table may be used to transport heavy equipment and to provide auxiliary bench space. With height adjustments from 28,75 to 35,75", it may be raised to the same height as most laboratory benchtops. Solid epoxy work surface, powder-coated square tubular steel frame.
  • Non-marking polyurethane castors
  • With toe locks