Transporting cart
Lieferant: LABCONCO
8019000
LBCP8019000EA 0 EUR
LBCP8019000
Transporting cart
Transportwagen
Tranporting cart made of stainless steel with two removable polished type 304 stainless steel pans that are 51 mm deep on three sides with one open side for easy equipment transfer from countertop to cart holds a variety of laboratory supplies, equipment and miscellaneous containers.