Glassware carts, used to carry glasswares, plasticwares and small instruments in vinyl-coated wire baskets. Choose from 2-baskets or 4-basket styles complete with removable plastic drip pan to catch spills.
Four-inch diameter casters absorb vibration over rough, uneven floors
Two toe-locks keeps the cart stationary when needed
Benches have 5" diameter, non-marking toe-locking casters with bearings for quiet operation