Trolleys, glasswares
Lieferant: LABCONCO
8032500
LBCP8032500EA 0 EUR
LBCP8032500
Trolleys, glasswares
Transportwagen
Glassware carts, used to carry glasswares, plasticwares and small instruments in vinyl-coated wire baskets. Choose from 2-baskets or 4-basket styles complete with removable plastic drip pan to catch spills.
  • Four-inch diameter casters absorb vibration over rough, uneven floors
  • Two toe-locks keeps the cart stationary when needed
  • Benches have 5" diameter, non-marking toe-locking casters with bearings for quiet operation