Eyewash bottle, sterilised, diphoterine, Esculape
Lieferant: LABORATOIRES ESCULAPE
8205020
LBES8205020EA 108 EUR
LBES8205020
Eyewash bottle, sterilised, diphoterine, Esculape
Erste-Hilfe-Ausstattung Augenduschen
Sterilised individual eye wash containing diphoterine.
  • Neutralises and evacuates an acid projection, basic or solvent
  • Sealed ensuring the integrity of the device
  • Disposable
Shelf life: Two years
Lieferumfang: Bottle is packed in a storage pouch with belt loop.