THERMOCYCLER GS4 OHNE BLÖCKE Manual Lid Temperature range: 4°C-99°C Temperature control: Calculated mode with plate and tube control algorithms Volume range: 2-150 µ l Block accuracy: +/- 0.4°C (20-99°C) Block homogeneity: +/- 0.4°C Ramp rate: Up to 1 * 1 ST
Lieferant: LABTECH
THERMOCYCLER GS4 OHNE BLÖCKE Manual Lid Temperature range: 4°C-99°C Temperature control: Calculated mode with plate and tube control algorithms Volume range: 2-150 µ l Block accuracy: +/- 0.4°C (20-99°C) Block homogeneity: +/- 0.4°C Ramp rate: Up to 1 * 1 ST