THERMAL CYCLER GS-4822 Specifications: Block materials: 2 x independent 48 well blocks: modular nickel plated anodised aluminium blocks with 4 Peltier heat modules and 2 thermistor sensors per block Traceability: NIST traceable temperature calibratio 1 * 1 ST
Lieferant: LABTECH
THERMAL CYCLER GS-4822 Specifications: Block materials: 2 x independent 48 well blocks: modular nickel plated anodised aluminium blocks with 4 Peltier heat modules and 2 thermistor sensors per block Traceability: NIST traceable temperature calibratio 1 * 1 ST