DAPI Counterstain (0.1ug/mL) 1mL LK095A DAPI Counterstain is used as a counterstain for chromosomes. The counterstain consists of DAPI and antifade, which helps preserving the fluorescence signal. DAPI finds strongly to FISH, particularly to AT rich 1 * 1 mL
Lieferant: LEICA MICROSYSTEMS
DAPI Counterstain (0.1ug/mL) 1mL LK095A DAPI Counterstain is used as a counterstain for chromosomes. The counterstain consists of DAPI and antifade, which helps preserving the fluorescence signal. DAPI finds strongly to FISH, particularly to AT rich 1 * 1 mL
Neuer Artikel bei VWR!