Stopcocks for burette, straight
Lieferant: Lenz Laborglas GmbH & CO.KG
02700501
LENZ02700501EA 27.2 EUR
LENZ02700501
Stopcocks for burette, straight
Hähne
NS stopcock made of borosilicate glass 3.3, facetted tip with plug of PTFE and screwthread retaining device.
  • Excellent chemical resistance
  • Excellent temperature resistance