Pharmaceutical Underbench Perfection Refrigerators Compliant with DIN 13277, HMTvh
Lieferant: Liebherr
HMTVH1501 HMTVH1501_H63 HMTVH1511 HMTVH1511_H63
LIEBHMTVH1501EA 1999 EUR
LIEBHMTVH1501 LIEBHMTVH1501_H63 LIEBHMTVH1511 LIEBHMTVH1511_H63
Pharmaceutical Underbench Perfection Refrigerators Compliant with DIN 13277, HMTvh
Kühlschränke
Ultra-efficient pharmaceutical refrigerators have been designed for the particular requirements with their dynamic cooling system, highly effective insulation, and precise temperature control, they guarantee reliable storage of sensitive substances and preparations.
  • Integrated data logger and USB interface for reading the data logger
  • PT1000 product sensor and access port for external probes
  • Min./max. temperature log shows whether the storage temperatures were within the tolerance limits
  • Insulated and glass door models with vertical LED lighting, self-closing door at <90° and electronic lock can be securely locked with a PIN code
  • Integrated LAN/WiFi and local API
  • Antimicrobial handle with lever opening mechanism
  • Underbench models: 819 mm external height, fits under most benches, worktop available if free-standing
The hygienic seamless moulded inner liner is made commercial-grade polystyrol. This makes it quick and easy to clean the large, rounded corners. Coated wire grid shelves guarantee constant air circulation for temperature stability. A pull-out stop prevents the shelf from sliding out when removing items.
Alarm relay and test function available. Alarm with battery backup (12 h) immediately upon power failure. Alarm history with real-time clock and 10 events and custom alarm settings possible. In the event of an uncontrolled temperature rise or other problems, the interior light alarm of the appliance starts flashing.
If the temperature falls below 2 °C due to a malfunction, the SafetyDevice takes control, stabilising the temperature, activating the alarm, and protecting the contents from freezing. ±2 K deviation from setpoint (H63 version with pharmacy drawers ±3 K), also with calibration option. A 12mm integrated access port allows a separate sensor to be connected for additional temperature monitoring without having to drill holes or break the door seal. Forced-air cooling system.
Pharmacy drawers with the practical system made of cold-conducting aluminium comes with a transparent front, perforated base, flexible dividing walls and a ¾-extension drawer. Medicines, vaccines and other items can be quickly found and the door closed again with minimal loss of cold air.
Features: Individually adjustable maintenance timer. Evaporator cover (prevents items from freezing when too close to the rear wall). Electronic door lock with remote control and SmartLock functionality. Replaceable door seal. SmartModule WiFi/LAN interface integrated. Steel/white external cabinet finish.
Standards: IEC 61010-2-011 standard refrigerators specially developed for storing temperature-sensitive products are CE safety-compliant. This ensures that all mechanical and electrical requirements for injury-free operation are met and that you and your team are always safe.