Perfection and Performance Laboratory Underbench Refrigerators, SRTvh and SRTvg
Lieferant: Liebherr
SRTVG1501 SRTVG1511 SRTVH1501 SRTVH1511
LIEBSRTVG1501EA 0 EUR
LIEBSRTVG1501 LIEBSRTVG1511 LIEBSRTVH1501 LIEBSRTVH1511
Perfection and Performance Laboratory Underbench Refrigerators, SRTvh and SRTvg
Kühlschränke
Glass/steel door underbench refrigerators with forced air cooling. White steel exterior and commercial-grade polystyrol interior. Choose between two temperature stability options to suit differing lab requirements. The intuitive user interfaces provide useful extended multi-functionality, e.g. a maintenance timer, and the LED interior lighting ensures good visibility of the contents of these appliances. Advanced connectivity options to suit varying requirements for simplified data storage, temperature control and alarm signals. These advanced lab fridges also feature a 2 °C safety device to protect sensitive contents from freezing.
  • Extended range of models for more choice to fulfil specific user requirements
  • Hygienic and long-lasting seamless polystyrol interior with a 12 mm access port for external probes
  • Energy-efficiency optimised appliances for lower running costs and help to meet user sustainability targets
  • 819 mm external height, fits under all common lab benches. Worktop available if free-standing
  • Antimicrobial handle with lever opening mechanism
  • Forced-air cooling system designed for very small internal temperature variation
Models are available in both perfection and performance series. Wire shelves with 45 kg shelf load, perforated sheet-metal shelves available as accessory for storing small items.
The choice of temperature stability, tested to EN 60068-3, important when it comes to validating appliances to your application or requirements. Perfection series ±2 K deviation from setpoint and DIN 13277 compliance. Performance series with ±3 K deviation from setpoint. Both with calibration function.
Perfection series for maximum content security: alarm with battery backup (12 h) immediately upon power failure, alarm history (10 events) and integrated data logger with real-time clock. Performance series: Power failure alarm when mains power returns.
Perfection series with integrated LAN/WiFi or local API for continuous monitoring and with USB interface for reading the data logger. Performance series with optional LAN/WiFi interfaces and local API (optional via SmartModule as accessory).
Standards: IEC 61010-2-011 standard refrigerators specially developed for storing temperature-sensitive products are CE safety-compliant. This ensures that all mechanical and electrical requirements for injury-free operation are met and that you and your team are always safe. Perfection series comply with DIN 13277.