Mueller Hinton Fastidious Agar
Lieferant: LIOFILCHEM
10132
LIOF10132EA 39.7 EUR
LIOF10132
Mueller Hinton Fastidious Agar
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
Mueller Hinton fastidious agar (Horse blood 5% + 20 mg/L β-NAD) is recommended by EUCAST for antimicrobial susceptibility testing of Streptococcus spp (including S. pneumoniae), Haemophilus spp and several other fastidious organisms.