Sterile Distilled Water
Lieferant: LIOFILCHEM
473030 463200 20097 21097
LIOF473030EA 49.8 EUR
LIOF473030 LIOF463200 LIOF20097 LIOF21097
Sterile Distilled Water
Medien für die Mikrobiologie Additive und Supplemente für die Mikrobiologie
Sterile distilled water is generally used for the reconstitution of certain products or preparation of the initial suspension of a sample and further dilutions.
Store at 10 to 25 °C away from light. Do not use the product beyond its expiry date on the label or if product shows any evidence of contamination or any sign of deterioration.
Standards: EN ISO 11133.
Bitte beachten For laboratory and professional use only.