Albert color kit Färbemittel für Bakteriologie
Lieferant: LIOFILCHEM
80294
LIOF80294EA 54.8 EUR
LIOF80294
Albert color kit Färbemittel für Bakteriologie
Albert color kit
Complete kit for Corynebacterium diphteriae staining according to Albert.
ALBERT COLOR KIT is a kit for staining diphtheria corynebacteria present in clinical samples or culture media. The stain reveals the granules of volutin present in Corynebacterium diphtheriae and other species of the genus Corynebacterium.
The reagents are containing in plastic bottles, sealed by thermo-induction and provided with a dropper top.
Each kit contains:
• 1 bottle containing 250 ml of toluidine blue – malachite green solution
• 1 bottle containing 250 ml of Lugol’s solution
• 1 Instruction sheet
IVDLogo