Albert color kit Färbemittel für Bakteriologie
Lieferant: LIOFILCHEM
Complete kit for Corynebacterium diphteriae staining according to Albert.
ALBERT COLOR KIT is a kit for staining diphtheria corynebacteria present in clinical samples or culture media. The stain reveals the granules of volutin present in Corynebacterium diphtheriae and other species of the genus Corynebacterium.
The reagents are containing in plastic bottles, sealed by thermo-induction and provided with a dropper top.
Each kit contains:
• 1 bottle containing 250 ml of toluidine blue – malachite green solution
• 1 bottle containing 250 ml of Lugol’s solution
• 1 Instruction sheet
![]() |