LiChrospher® RP-18, HPLC Columns
Lieferant: Avantor
LISPRP18-5-50AFEA 784 EUR
LISPRP18-5-50AF LISPRP18-5-10C5 LISPRP18-5125A LISPRP18-5-125AF LISPRP18-5-150AF LISPRP18-5250A LISPRP18-5-250AF LISPRP18-5-250AM LISPRP185300A LISPRP185300AF LISPRP18575AF LISPRP18510C5S
LiChrospher® RP-18, HPLC Columns
Chromatographiesäulen
These LiChrospher® columns are silica available both endcapped and non-endcapped C18 phases. This feature enhances the chromatography selectivity options.
  • Spherical porous silica
  • High efficiency packed columns