DNA ladders, SimplyLoad™
Lieferant: LONZA
50327 50333
LONZ50327EA 235 EUR
LONZ50327 LONZ50333
DNA ladders, SimplyLoad™
Elektrophorese-Reagenzien Molekulargewichtsmarker
Lonza SimplyLoad™ DNA ladders are ready to load directly onto the gel to maximise time and convenience. Each band has an ideal 50% G-C content for rapid, precise sizing and sharp resolution. Free from contaminating plasmid DNA so they have minimal background.
  • Ready to load
  • Storage temperature: 2 to 8 °C