DNA QuantLadders
Lieferant: LONZA
50334
LONZ50334EA 216 EUR
LONZ50334
DNA QuantLadders
Elektrophorese-Reagenzien Molekulargewichtsmarker
QuantLadders are prepared from five highly purified DNA fragments that provide visual quantitation of DNA within ±1% accuracy. The DNA QuantLadder provides each DNA fragment in perfect relative mass equivalence at 100 ng, 70 ng, 50 ng, 20 ng and 10 ng. The reverse QuantLadder simply reverses the mass equivalence order of the QuantLadder.
  • Storage temperature: 2 to 8 °C
  • Also available in SimplyLoad® format