gelStar™ Nucleic acid gel stain 10,000X
Lieferant: LONZA
50535
LONZ50535EA 344 EUR
LONZ50535
gelStar™ Nucleic acid gel stain 10,000X
Färbemittel Elektrophorese-Färbemittel
GelStar is a sensitive fluorescent stain for detecting both DNA and RNA.
  • Maximum sensitivity - detect as little as 20 pg of dsDNA or 3 ng of RNA,
  • Versatile - use for agarose or polyacrylamide gel electrophoresis,
  • Ideal alternative to silver staining
GelStar™ Nucleic Acid Gel Stain is a highly sensitive fluorescent stain for detecting both DNA and RNA. You can choose to either add GelStarTM Stain to your agarose solution prior to casting or to post stain your gels. GelStarTM Nucleic Acid Gel Stain exhibits exceptional signal-to-noise ratio with minimal background. Maximum sensitivity, detect as little as 20 pg of dsDNA or 3 ng of RNA