Protein markers, ProSieve™ Colour
Lieferant: LONZA
50550 50552
LONZ50550EA 231 EUR
LONZ50550 LONZ50552
Protein markers, ProSieve™ Colour
Elektrophorese-Reagenzien Molekulargewichtsmarker
ProSieve™ Color protein marker have been labelled with three different fluorescent dyes. These markers are supplied as a lyophilised solid containing the buffer salts and detergent found in the typical Laemmli buffer system. It contains nine proteins with approximate masses of 10, 15, 20, 25, 40, 50, 80, 125, and 190 kDa.
  • Ideal for use as visible marker in SDS-PAGE gels and Western blotting
  • Individually calibrated against a ProSieve™ unstained protein marker