SYPRO® Ruby protein blot stain
Lieferant: LONZA
50565
LONZ50565EA 311 EUR
LONZ50565
SYPRO® Ruby protein blot stain
Färbemittel Elektrophorese-Färbemittel
SYPRO™ Ruby is 60-times more sensitive than reversible protein blot stains like Ponceau S, and 30-times more sensitive than Amido Black or Coomassie® brilliant blue stains.
  • Visualize proteins prior to transfer - does not interfere with protein activity
  • Versatile - Excellent for staining native proteins
  • Safe - No acids or organic solvents necessary
  • Sensitive - Detects as little as 4-8 ng protein per band