SYPRO® Ruby protein blot stain
Lieferant: LONZA
50565
LONZ50565EA
311
EUR
LONZ50565
SYPRO® Ruby protein blot stain
Färbemittel
Elektrophorese-Färbemittel
SYPRO™ Ruby is 60-times more sensitive than reversible protein blot stains like Ponceau S, and 30-times more sensitive than Amido Black or Coomassie® brilliant blue stains.
- Visualize proteins prior to transfer - does not interfere with protein activity
- Versatile - Excellent for staining native proteins
- Safe - No acids or organic solvents necessary
- Sensitive - Detects as little as 4-8 ng protein per band