ORP meter, handheld, SD 60
Lieferant: Lovibond
194801
LOVI194801EA 166 EUR
LOVI194801
ORP meter, handheld, SD 60
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
The SD 60 ORP meter is a compact, easy to use meter for the measurement of OPR or redox potential.
  • Intuitive scroll-through functionality
  • Robust plastic case with hanger
  • Data storage function for 25 sets of data, each with date and time stamp
  • Backlit display
With robust housing and fully waterproof (IP67) casing, this tester is the ideal solution for in-situ testing in environmental, industrial or pool & spa applications. The series can be upgraded with replaceable electrodes to ensure long-life functionality in the field.
Lieferumfang: Meter is supplied with two batteries, lanyard, robust carrying case and instruction manual.