Reference standard kits for use with MD 100 and MD 200 photometers
Lieferant: Lovibond
205600 205610 205640 205605 275660 275655 205606 275650 275656 275670
LOVI205600EA 85.6 EUR
LOVI205600 LOVI205610 LOVI205640 LOVI205605 LOVI275660 LOVI275655 LOVI205606 LOVI275650 LOVI275656 LOVI275670
Reference standard kits for use with MD 100 and MD 200 photometers
Standards Umweltanalyse-Standards
Reference standards are designed to check the accuracy and the reliability of the results.
  • With a shelf life of two years
  • Not possible to calibrate the photometer with the reference standards