Reference standard kits for use with MD 100 and MD 200 photometers
Lieferant: Lovibond
205600
205610
205640
205605
275660
275655
205606
275650
275656
275670
LOVI205600EA
85.6
EUR
LOVI205600
LOVI205610
LOVI205640
LOVI205605
LOVI275660
LOVI275655
LOVI205606
LOVI275650
LOVI275656
LOVI275670
Reference standard kits for use with MD 100 and MD 200 photometers
Standards
Umweltanalyse-Standards
Reference standards are designed to check the accuracy and the reliability of the results.
- With a shelf life of two years
- Not possible to calibrate the photometer with the reference standards