Suspended solids colorimeter, MD100
Lieferant: Lovibond
The MD 100 photometer is a portable handheld instrument with a simple button layout, robust waterproof design and large backlit display that are ideal for testing in the field or laboratory conditions.
- Accurate (3% FS), reproducible results using high quality interference filter with long-life LED
- Internal ring memory for 16 data sets
- Real time clock and date
- OTZ (One Time Zero setting) on versions except those denoted*
- Optional infrared interface for transfer of test data to PC or printer
Lieferumfang: Instrument supplied with carry case, 4× AAA batteries, 3× round glass vials with lids, relevant reagents, and instruction manual.