Test kits, arsenic
Lieferant: Lovibond
400700
LOVI400700EA 187 EUR
LOVI400700
Test kits, arsenic
Umwelt-Prüfkits
High sensitivity kit suitable for the determination of arsenic in drinking water.
  • Detects 0,005 mg/l arsenic
  • A solid acid substance is used in order to avoid any irritation by a corrosive acid on the user’s hands
  • The unbreakable plastic reaction vessel is more convenient and safe for on-site testing
Lieferumfang: The test kit contains a water-proof colour chart which also includes the brief instruction for use in pictograms. Supplied in a carrying case.