Test kits, sodium hypochlorite, Comparator 2000+
Lieferant: Lovibond
411390
LOVI411390EA 692.3 EUR
LOVI411390
Test kits, sodium hypochlorite, Comparator 2000+
Umwelt-Prüfkits
  • Stable and accurate
  • Robust, compact and portable
  • Prism enabling colour matching
  • Diffuser plate is used to measure the turbidity
  • Longer path length cells be used for lower concentration analysis
The integrated prism brings the glass standards of the test discs and the coloured sample into the same field of view.
Lieferumfang: Kit is supplied in a robust carrying case containing Comparator 2000+, sodium hypochlorite comparator disc, 2× comparator vials, 10 ml, 13 mm path length, tablet reagents, crushing / stirring rod, 2× shaker tubes, plastic syringes 1 and 5 ml, vial cleaning brush and instruction sheet.