Methylrot aqueous solution

Lieferant: Lovibond

Synonyme: 2-(4-(Dimethylamino)phenylazo)benzoesäure

451630 451631
LOVI451630EA 18.1 EUR
LOVI451630 LOVI451631
Methylrot aqueous solution
Methylrot
Formel: C₁₅H₁₅N₃O₂
Molecular Weight: 269.3 g/mol
MDL: MFCD00002425
CAS-Nummer: 493-52-7
EINECS: 207-776-1
Merck Index: 12,06199

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR