Neßler-Reagenz

Lieferant: Lovibond
Gefahr

Synonyme: Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution

465200 465201
LOVI465200EA 23.1 EUR
LOVI465200 LOVI465201
Neßler-Reagenz
Neßler-Reagenz
Siedepunkt: ∼100 °C (1013 hPa)
Dichte: 1,2 g/cm³ (20 °C)
MDL: MFCD00049656
CAS-Nummer: 7783-33-7
UN: 2922
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR