Neßler-Reagenz, Tabletten

Lieferant: Lovibond

Synonyme: Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution

511261BT 511231BT 511230BT 511251BT 511250BT
LOVI511261BTEA 123.8 EUR
LOVI511261BT LOVI511231BT LOVI511230BT LOVI511251BT LOVI511250BT
Neßler-Reagenz, Tabletten
Neßler-Reagenz
MDL: MFCD00049656
CAS-Nummer: 7783-33-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR