Buffer tablets, Lovibond®

Lieferant: Lovibond

515621BT 515620BT 515600BT 515601BT 515610BT 515611BT
LOVI515621BTEA 51.9 EUR
LOVI515621BT LOVI515620BT LOVI515600BT LOVI515601BT LOVI515610BT LOVI515611BT
Buffer tablets, Lovibond®
Puffer pH-Puffer

Ideal for preparing buffer solutions of 4, 7 and 10 pH units.


pH tablet can be added to 100 ml of distilled or deionised water for preparing solution of specified concentration.

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR