Transport container, Crioplast®
Lieferant: LP ITALIANA
100015_U
LPIT100015_UEA 54.2 EUR
LPIT100015_U
Transport container, Crioplast®
Versandtaschen und Transportbehälter
Plastic, white, with blue inserts.
Crioplast® refrigerated container for maintaining and transporting blood at original conditions.