Microcentrifuge tubes, 1,5 ml
Lieferant: LP ITALIANA
111540
LPIT111540EA 350 EUR
LPIT111540
Microcentrifuge tubes, 1,5 ml
Röhrchen Reaktionsgefäße
PP, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Graduated.
With flat cap.