Test tube racks, ABS
Lieferant: LP ITALIANA
181082 182082
LPIT181082EA 276 EUR
LPIT181082 LPIT182082
Test tube racks, ABS
Gestelle Reagenzglasgestelle
Stackable and non floating ABS rack with alphanumeric grid.
  • Good temperature resistance
  • Good chemical resistance