Containers, wide mouth, with screw cap
Lieferant: LP ITALIANA
190100 190015 190200
LPIT190100EA 163 EUR
LPIT190100 LPIT190015 LPIT190200
Containers, wide mouth, with screw cap
Probenbehälter Universalbehälter
Container made of PS, clear, with white screw cap.
  • Good chemical resistance
  • Lightweight
  • Easy to clean
Suitable for surgical or anatomical specimens.