Stoppers, butyl, for diagnostic vials, WHEATON®
Lieferant: DWK Life Sciences
600005
LSUK600005EA 159 EUR
LSUK600005
Stoppers, butyl, for diagnostic vials, WHEATON®
Stopfen Gummistopfen
Stoppers are made from the pharmaceutical grade materials.
  • Suitable for freeze drying