Capsules for microbalances
Lieferant: LUDI
176981126
LUDI176981126EA 160 EUR
LUDI176981126
Capsules for microbalances
Gewichte Kalibriergewichte
Made of silver.
  • Very good chemical resistance
  • Very good temperature resistance
They can be used with all commercially available analysers and microbalances.