Drum pumps B2 Vario PP
Lieferant: LUTZ PUMPEN
0205-022 0201-502 0205-020 0205-021 0201-501 0201-500
223-0087EA 854 EUR
223-0087 LUTZ0201-502 223-0085 223-0086 LUTZ0201-501 LUTZ0201-500
Drum pumps B2 Vario PP
Pumpen Fass/Transferpumpen
Drum pumps for thin bodied fluids such as battery acids, amonia solutions, photographic developer/-fixer, glycol, phosphoric acids, hydrochloric acid and hydorgen peroxides.For filling small quantities from hobbocks, canisters and drums
  • Due to the modular construction, the pump is service-friendly
  • The pumps offer a complete liquid drainage function
  • Already pre-assembled: immediately ready for action
  • Ideal for pumping and filling thin bodied liquids
Bestellinformation: Available as a set with accessories or pump-only (no hose or other accessories).