Drum pumps sets for mineral oils
Lieferant: LUTZ PUMPEN
0205-301
LUTZ0205-301EA 1180 EUR
LUTZ0205-301
Drum pumps sets for mineral oils
Pumpen Fass/Transferpumpen
Ideal for light viscous mineral oil products.
Lieferumfang: Pump set consists of one motor MI 4, pump tube Alu 41-L-SL, hose connenction Alu 1", 2 m PVC fabric hose 1", two hose clips 1", nozzle aluminium, hose connectors brass 1" and drum adapter PP.