Mycophenolic acid-O-β-D-glucuronide-Lösung Acetonitrile:Water (90:10) (v/v) 1 mg/ml, Cerilliant®, Zertifiziertes Referenzmaterial

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ300

M-250-1ML
M-250-1MLEA 301 EUR
M-250-1ML
Mycophenolic acid-O-β-D-glucuronide-Lösung Acetonitrile:Water (90:10) (v/v) 1 mg/ml, Cerilliant®, Zertifiziertes Referenzmaterial
Mycophenolic acid-O-β-D-glucuronide

Mycophenolic acid (MPA), the active moiety of the prodrug mycophenolate mofetil, is widely used in immunosuppressive regimens after kidney, liver or heart transplantation. MPA is metabolized predominantly to the inactive 7-O-glucuronide (MPAG). A minor fraction is converted to the pharmacologically active acyl glucuronide (AcMPAG). All compounds ultimately are eliminated via the kidneys.

Formel: C₂₃H₂₈O₁₂
Flammpunkt: 2,0 °C (geschlossener Tiegel)
Lagertemperatur: Gefrierschrank
MDL: MFCD09878308
CAS-Nummer: 31528-44-6
UN: 1648
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR