N-Methylbistrifluoracetamid, Supelco®

Lieferant: Merck
Gefahr

Synonyme: 2,2,2-Trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide, MBTFA

M-Clarity™ Qualitätsstufe = MQ100

M0789-10X1ML M0789-5ML
M0789-10X1MLEA 532 EUR
M0789-10X1ML M0789-5ML
N-Methylbistrifluoracetamid, Supelco®
N-Methylbistrifluoracetamid
Formel: C₅H₃F₆NO₂
Molecular Weight: 223.07 g/mol
MDL: MFCD00000412
CAS-Nummer: 685-27-8
EINECS: 211-680-5
UN: 2920
ADR: 8.3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79168343a158a1d2fad009b16d47996ff4ace6b4&vid=137483508520ebf407de9eff5c9f28766216fb7a&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2aa16dc54546c9e52c7956ea07e20f550635839&vid=fb0a343c7442e32e05ee393fb59950ff80594016&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR