Acrylamid 40% aqueous solution, Acryl 40™, Proteomics grade
Lieferant: VWR Chemicals
|
Gefahr
|
Synonyme:
Propenamid, Acrylsäureamid, Propensäureamid, 2-Propenamid
VWR offers an extensive line of acrylamide and bis-acrylamide pre-weighed powder blends, pre-mixed stock solutions, and ready to use solutions for customised polyacrylamide gel electrophoresis of proteins and PAGE of nucleic acids. Our ultra-pure (>99,9%) acrylamide and bis-acrylamide powders provide the flexibility to prepare solutions having concentrations and ratios for all electrophoresis applications. Liquid blends minimise the handling of neurotoxic acrylamide.
- Manufactured from ultra-pure powders with acrylamide purity >99,9%
- Convenient, ready to use
- Eliminates hazardous powdered acrylamide handling
Formel:
C₃H₅NO Molecular Weight: 71.08 g/mol Siedepunkt: 232 Schmelzpunkt: 84.5 Flammpunkt: 138 Lagertemperatur: Kühlschrank |
MDL:
MFCD00008032 CAS-Nummer: 79-06-1 EINECS: 201-173-7 UN: 3426 ADR: 6.1 III |
|
Spezifikation Testergebnisse
Acrylsäure | < 0,005% |
Leitfähigkeit | < 5 umhos |
DNasen | Keine nachgewiesen |
Elektrophorese | Entspricht |
pH@25°C | 6 - 7 |
Proteasen | Keine nachgewiesen |
RNasen | Keine nachgewiesen |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...