Harnstoff 8 mol/l in wässriger Lösung, Proteomik-Qualität

Lieferant: VWR Chemicals

Synonyme: Urea, Aminoameisensäureamid, Carbamid, Carbonyldiamid

M124-250MLEA 173 EUR
M124-250ML
Harnstoff 8 mol/l in wässriger Lösung, Proteomik-Qualität
Harnstoff

A convenient pre-mixed solution that is easily diluted for buffer preparation under denaturing conditions.Commonly used to denature nucleic acids for electrophoresis and to study the secondary and tertiary structure of proteins. Effective for maintaining DNA in a highly denatured state during sequencing protocols, urea is also useful for accelerating the activity of Proteinase K digestions.

Formel: NH₂CONH₂
Molecular Weight: 60.06 g/mol
Siedepunkt: 197
Schmelzpunkt: 132,7
Lagertemperatur: Raumtemperatur
MDL: MFCD00008022
CAS-Nummer: 57-13-6
EINECS: 200-315-5
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Clarity Pass
Protease none detected
Urea 7,8 - 8,2 Molar
Appearance Clear and colorless liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR