Next generation sequencing (NGS), DNA/RNA clean up Mag-Bind® TotalPure
Lieferant: OMEGA BIO-TEK
M1378-00 M1378-01 M1378-02
M1378-00EA 134 EUR
M1378-00 M1378-01 M1378-02
Next generation sequencing (NGS), DNA/RNA clean up Mag-Bind® TotalPure
Nukleinsäure-Reagenzien Next Generation Sequencing Reagenzien
Mag-Bind® TotalPure NGS offers an easy-to-use, reliable solution for the purification of both DNA and RNA for next-generation sequencing workflows with high recovery rates. Mag-Bind® TotalPure NGS is capable of selectively binding fragments depending on the reagent to sample ratio used, giving the user flexibility to perform left, right or double-sided size selection.
  • One stop solution for both DNA and RNA clean-up
  • Double-sided size selection capability
  • No centrifugation steps
  • Manual and automation-friendly sample processing in 96- and 384-well formats
Mag-Bind® TotalPure NGS is designed for both manual and fully automated purification of DNA and RNA samples, as well as for the purification of PCR products. The system combines proprietary chemistries with the reversible nucleic acid-binding properties of magnetic beads to selectively bind fragments larger than 100 bp and eliminate excess nucleotides, primers, and small, non-targeted products, such as primer-dimers. Purified DNA and RNA is suitable for a variety of downstream applications, such as NGS library preparation, microarrays, automated fluorescent sequencing and restriction enzyme digestion.