Benzamidin Hydrochlorid Hydrat ≥98,0% (durch Trockensubstanz), Proteomik-Qualität
Lieferant: VWR Chemicals
|
Achtung
|
A protease inhibitor of arginine-selective enzymes like trypsin, kallikrein and thrombin.
- Product is animal-free
Extracted proteins are exposed to the threat of degradation by proteases from the moment cells or tissues are lysed. Proteases present in the living cell are released from their regulated environment into the extract where they may freely degrade their targets, resulting in drastically reduced protein yield and sample
quality. Complex protein solutions are typically treated with a mixture of several protease inhibitors in order to preserve the native state of the proteins.
Formel:
C₆H₅C(=NH)NH₂·HCl·H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066285 CAS-Nummer: 206752-36-5 EINECS: 216-795-4 |
Spezifikation Testergebnisse
molarer Extinktionskoeffizient (Lambda Max, Methanol, wasserfrei) | |
Schwermetalle (wie Pb) | ≤ 0,005 % |
Identität (IR) | entspricht |
Schmelzpunkt | 77-87 °C |
Reinheit (bez. auf wasserfrei) | ≥ 98,0% |
Datum der nochmaligen Untersuchung | siehe Analysenzertifikat |
Löslichkeit (1 %, Wasser) | entspricht |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...