Chromazurol B
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
Mordant Blue 1, Eriochrome Azurol B, Sunchromine Pure Blue BX
M-Clarity™ Qualitätsstufe = MQ200
M1782-1G
M1782-1GEA
368
EUR
M1782-1G
Chromazurol B
Chromazurol B
Chromeazurol B is a dye, belonging to the class of triphenylmethanes.
- Chromeazurol B may be used as a synthetic dye in the determination of trace levels of uranium in water samples using spectrophotometric technique
Formel:
C₂₃H₁₄Cl₂Na₂O₆ Molecular Weight: 503,24 g/mol Lagertemperatur: Room Temperature |
MDL:
MFCD00036446 CAS-Nummer: 1796-92-5 |
Spezifikation Testergebnisse
Appearance | Brown with an orange to red cast powder |
Solubility | Slightly hazy orange solution at 4 mg plus 4 ml of water |
Dye content | approx. 60% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...