Molybdenum Blue reagent, Sprühreagenz, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

M1942-100ML
M1942-100MLEA 111 EUR
M1942-100ML
Molybdenum Blue reagent, Sprühreagenz, Sigma-Aldrich®
Molybdenum Blue reagent

For use in the detection of phospholipids and related compounds.

Lagertemperatur: Raumtemperatur
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR