VWR® Separating Gel Buffer, 4X
Lieferant: VWR Chemicals
M197-500MLEA 375 EUR
M197-500ML
VWR® Separating Gel Buffer, 4X
Puffer Elektrophorese-Puffer Trennpuffer
4X concentrated buffer solution (1,5 M Tris, pH 8,8 and 0,4% SDS). Used for preparation of standard acrylamide:bis-acrylamide gels as well as preparation of gels using PAGE-PLUS™ acrylamide solution.