TRITON™ X-100 (Polyethylenglycol-tert-octylphenylether) 10% in wässriger Lösung, Proteomik-Qualität peroxidfrei
Lieferant: VWR Chemicals
|
Gefahr
|
M236-10ML-5PKEA
259
EUR
M236-10ML-5PK
TRITON™ X-100 (Polyethylenglycol-tert-octylphenylether) 10% in wässriger Lösung, Proteomik-Qualität peroxidfrei
TRITON™ X-100 (Polyethylenglycol-tert-octylphenylether)
High purity Pro-Pure™ grade detergents are carefully packed under inert gas, making them extremely stable.
- Optimised for isolating proteins and sample preparation
- Contains <1 ug/ml of peroxide
- Convenient concentrations pre-measured in ampoules
- Long-term stability
Lieferumfang: Enthält 5*10 ml Vials einer 10% igen Lösung.
Siedepunkt:
~100 °C (1013 hPa) Dichte: 1,01 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00132505 CAS-Nummer: 9002-93-1 |
Spezifikation Testergebnisse
Colour (APHA) | ≤100 |
Peroxides-free | Pass/fail |
pH (50%, Water, 25° C) | 6,0 - 8,0 |
Protease- free | Pass/fail |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...