Mag-Bind® Blood DNA HV Kit
Lieferant: OMEGA BIO-TEK
Isolate DNA from up to 10 ml blood using magnetic beads.
- DNA from up to 10 ml whole blood and saliva
The Mag-Bind® Blood DNA HV kit is designed for rapid and reliable isolation of high-quality genomic DNA from up to 10 ml whole blood and saliva. The protocol has been successfully automated on the Hamilton Microlab® STAR™ and 24 samples can be processed in less than 2,5 hours.
The system combines the reversible nucleic acid-binding properties of Mag-Bind® paramagnetic particles with the time-proved efficiency of Omega Bio-tek’s DNA isolation system to provide a fast and convenient method to isolate DNA from a variety of samples. The Mag-Bind® particles provide fast response time when applied to a magnetic field, allowing for quick separation from high volumes of liquid.